Learn More
Azurocidin/CAP37/HBP Antibody (308), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89593FR
Product Code. 18468655
  • € 625.00 / 0.10mL
Description
Azurocidin/CAP37/HBP Monoclonal antibody specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Azurocidin/CAP37/HBP
Monoclonal
DyLight 680
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA
308
ELISA
AZAMP, AZU, azurocidin, azurocidin 1, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin
This antibody was obtained from a rabbit immunized with purified, recombinant Human Azurocidin/CAP37/HBP (NP_001691.1; Met1-Pro250).
0.1 mL
Apoptosis
566
Store at 4°C in the dark.