Learn More
Brady™ B-483A Polyester Ultra Aggressive Thermal Transfer Labels
Product Code. 11508304
Click to view available options
Height (Metric):
12.7 mm
25.4 mm
50.8 mm
76.2 mm
Labels per Row:
1
2
3
Vertical Repeat (Metric):
15.2 mm
28.6 mm
54 mm
79.4 mm
Width (Metric):
25.4 mm
38.1 mm
76.2 mm
101.6 mm
Unit Size:
1 roll
Product Code. 11508304
Brand: Brady™ 031335
Only null left
Add to basket
Add to basket
Printable, ultra aggressive polyester labels for Thermal Transfer Printers.
  • 76 mm core
  • For smooth or textured surfaces
  • Aggressive, permanent adhesive
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance UL and CSA Approved
Adhesive Type Permanent Rubber
Color White
Finish Gloss
Format Die-cut
Material Polyester (B-483A)
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset Tracking, Equipment Labeling, Data, Telecommunications and Electrical Labeling, General Identification, Inventory Labeling
For Use With (Equipment) BBP81;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500
Horizontal Repeat (Metric) 41.9 mm
Labels per Row 2
Min. Operating Temperature (Metric) -40°C
Model THT-37-483-10
No. per Pack 10000 Label(s) / Roll
Quantity 10,000 Labels
Shape Rectangle
Size 38.10 mm (W) x 12.70 mm (H)
Temperature Range (English) -40°F to 248°F
Temperature Range (Metric) -40°C to +120°C
Thickness (English) 0.004 in
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 15.2 mm
Dimensions (LxW) 38.10 mm (W) x 12.70 mm (H)
Width (English) 1.5 in
Width (Metric) 38.1 mm
Width (English) Printable Area 38.10 mm
Width (Metric) Printable Area 38.1 mm
Height (English) 0.5 in
Height (Metric) 12.7 mm
Height (Metric) Printable Area 12.7 mm
Alternate Part Number THT-37-483-10