Learn More
Brady™ B-7610 Paper Thermal Transfer Labels, 76 mm Core
Product Code. 11569883
Change view
Click to view available options
For Use With (Equipment):
BBP11, BBP12, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
BBP11, BBP12, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Height (Metric):
50 mm
55.9 mm
80 mm
159 mm
Width (Metric):
80 mm
104 mm
108 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Height (Metric) Width (Metric) unitSize
11569883 BBP11, BBP12, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 50 mm 80 mm Each
11568774 BBP11, BBP12, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 55.9 mm 104 mm Each
11518784 BBP11, BBP12, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 80 mm 108 mm Each
11598774 BBP11, BBP12, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 159 mm 108 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11569883 Supplier Brady™ Supplier No. 620366
Stock Up Your Lab
Use promo code 28007
View all promotions
In Stock
Add to basket
Add to basket
Thermal transfer printable paper for low cost general labeling applications.
  • 76 mm core
  • Permanent acrylic adhesive
  • Matte finish
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Paper (B-7610)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification
For Use With (Equipment) BBP11, BBP12, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
Model THT-80x50-7610
No. per Pack 6000 Label(s) / Box
Quantity 6000 Labels
Shape Rectangle
Size 80.00 mm (W) x 50.00 mm (H)
Temperature Range (English) -40°F to 122°F
Temperature Range (Metric) -40°C to +50°C
Thickness (English) 0.0032 in.
Thickness (Metric) 0.081 mm
Dimensions (L x W) 80.00 mm (W) x 50.00 mm (H)
Length (English) 1.97 in.
Width (English) 3.15 in.
Width (Metric) 80 mm
Width (English) Printable Area 80.00 mm
Height (English) 1.97 in.
Height (Metric) 50 mm
Alternate Part Number THT-80x50-7610