missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-Raf Antibody (OTI5A9), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70260FR
This item is not returnable.
View return policy
Description
B-Raf Monoclonal specifically detects B-Raf in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| B-Raf | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| BRAF | |
| Full length recombinant protein of human B-Raf (NP_004324) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI5A9 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| 94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Phospho Specific | |
| 673.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction