missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B-Raf (V600E) Recombinant Rabbit Monoclonal Antibody (RM8), Biotin

Description
This antibody reacts to the Histone H3.3 G34R mutant. No cross reactivity with wild type Histone H3.3.
This gene, B-RAF/BRAF, encodes a protein belonging to the RAF family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERK signaling pathway, which affects cell division, differentiation, and secretion. Mutations in this gene, most commonly the V600E mutation, are the most frequently identified cancer-causing mutations in melanoma, and have been identified in various other cancers as well, including non-Hodgkin lymphoma, colorectal cancer, thyroid carcinoma, non-small cell lung carcinoma, hairy cell leukemia and adenocarcinoma of lung. Mutations in this gene are also associated with cardiofaciocutaneous, Noonan, and Costello syndromes, which exhibit overlapping phenotypes. A pseudogene of this gene has been identified on the X chromosome.
Specifications
Specifications
| Antigen | B-Raf (V600E) |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | RM8 |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide |
| Gene | BRAF |
| Gene Accession No. | P15056 |
| Gene Symbols | BRAF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?