missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ C-Flex™ 50-A B/T™ PerfectPosition™ Pump Tubing

Brand:  Masterflex™ 06424-90

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11891465

  • € 166.00 / Each


Estimated Shipment
03-08-2021
Add to basket

Description

Description

  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Heat sealable and weldable
  • Inexpensive
  • Biocompatible
  • Physical properties similar to silicone with chemical compatibility of Tygon™
Specifications

Specifications

FDA approved; Prop 65 NA (no Prop 65 chemicals)
2.4 bar
Yes
7.6 m
Sterile
42000 mL/min
135°C
660 mm Hg
C-Flex 50-A
-40°C
90 (19 mm)
Autoclave, Ethylene Oxide, Gamma Irradiation
Videos
SDS
Documents