missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B220/CD45R Antibody (IVA103), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-492APC
This item is not returnable.
View return policy
Description
B220/CD45R Monoclonal specifically detects B220/CD45R in Bovine samples. It is validated for Flow Cytometry.
Specifications
| B220/CD45R | |
| Monoclonal | |
| APC | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody IVA103 reacts with CD45R antigen (bovine), a single chain type I transmembrane glycoprotein (member of the protein tyrosine phosphatase family), a restricted form of the CD45 family. CD45R is an essential regulator of T and B lymphocytes antigen receptor signaling and it is specifically expressed in hematopoietic cells. | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| IVA103 | |
| Flow Cytometry | |
| PTPRC | |
| Bovine leukocytes. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Bovine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction