missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B220/CD45R Antibody (IVA103), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-492H
This item is not returnable.
View return policy
Description
B220/CD45R Monoclonal antibody specifically detects B220/CD45R in Bovine samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF
Specifications
| B220/CD45R | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
| IVA103 | |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF-ready | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Bovine leukocytes. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction