missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GALNT1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92677-0.02ml
This item is not returnable.
View return policy
Description
B3GALNT1 Polyclonal antibody specifically detects B3GALNT1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| B3GALNT1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| b3Gal-T3, Beta-1,3-galactosyltransferase 3, Beta-1,3-GalNAc-T1, Beta-1,3-GalTase 3, beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group), Beta3GalT3, Beta3Gal-T3, beta-3-Gx-T3, brainiac1, EC 2.4.1.79, Galactosylgalactosylglucosylceramide beta-D-acetyl-galactosaminyltransferase, Gb4Cer, GLCT3, Globoside synthase, globotriaosylceramide 3-beta-N-acetylgalactosaminyltransferase, P, P antigen synthase, P blood group globoside, polypeptide 3 (Globosideblood group), UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 1, UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 1(Globoside blood group), UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 1(Globoside blood group), UDP-N-acetylgalactosamine:globotriaosylceramidebeta-1,3-N-acetylgalactosaminyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 44-331 of human B3GALNT1 (NP_003772.1). IERVNWMYFYEYEPIYRQDFHFTLREHSNCSHQNPFLVILVTSHPSDVKARQAIRVTWGEKKSWWGYEVLTFFLLGQEAEKEDKMLALSLEDEHLLYGDIIRQDFLDTYNNLTLKTIMAFRWVTEFCPNAKYVMKTDTDVFINTGNLVKYLLNLNHSEKFFTGYPLIDNYSYRGFYQKTHISYQEYPFKVFPPYCSGLGYIMSRDLVPRIYEMMGHVKPIKFEDVYVGICLNLLKVNIHIPEDTNLFFLYRIHLDVCQLRRVIAAHGFSSKEIITFWQVMLRNTTCHY | |
| 0.02 mL | |
| Cardiovascular Biology | |
| 8706 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering