missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GALNT2 Antibody (OTI1G2), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72411IR
This item is not returnable.
View return policy
Description
B3GALNT2 Monoclonal specifically detects B3GALNT2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| B3GALNT2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| B3GALNT2 | |
| Full length recombinant protein of human B3GALNT2 (NP_689703) produced in HEK293T cells. | |
| 0.1 mL | |
| 148789.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1G2 | |
| Western Blot | |
| B3GalNAc-T2, beta 1,3-N-acetylgalactosaminyltransferase-II (MGC39558), beta-1,3-galactosaminyltransferase 2, Beta-1,3-GalNAc-T2, beta-1,3-N-acetylgalactosaminyltransferase 2, Beta-1,3-N-acetylgalactosaminyltransferase II, EC 2.4.1, EC 2.4.1.-, MGC39558, UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 2, UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 2, UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction