Learn More
B3GALNT2 Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505973S
This item is not returnable.
View return policy
Description
B3GALNT2 Monoclonal antibody specifically detects B3GALNT2 in Human samples. It is validated for Western BlotSpecifications
| B3GALNT2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P78358 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1485 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1G2 | |
| Unconjugated | |
| CTAG1B | |
| A930105D20Rik; B3GalNAc-T2; B3galnt2; Beta-1,3-GalNAc-T2; beta-1,3-N-acetylgalactosaminyltransferase 2; beta-1,3-N-acetylgalactosaminyltransferase II; C80633; D230016N13Rik; MDDGA11; RGD1306946; UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 2; UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 2; UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 2 | |
| Full length human recombinant protein of human CTAG1B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |