Learn More
B3GALNT2 Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505973
This item is not returnable.
View return policy
Description
Beta-1,3-N-acetylgalactosaminyltransferase active in synthesizing a unique carbohydrate structure, GalNAc-beta-1-3GlcNAc, on N- and O-glycans. Has no galactose nor galactosaminyl transferase activity toward any acceptor substrate.Specifications
| B3GALNT2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NCR0 | |
| B3GALNT2 | |
| Full length human recombit protein of human B3GALNT2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1G2 | |
| Unconjugated | |
| B3GALNT2 | |
| B3GalNAc-T2, MDDGA11 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 148789 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |