Learn More
B3GALNT2 Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505973
Encompass_Preferred
 View more versions of this product
Product Code. 17575162
  • € 540.87 / 100µL
Description
Beta-1,3-N-acetylgalactosaminyltransferase active in synthesizing a unique carbohydrate structure, GalNAc-beta-1-3GlcNAc, on N- and O-glycans. Has no galactose nor galactosaminyl transferase activity toward any acceptor substrate.
Specifications
Specifications
B3GALNT2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8NCR0
B3GALNT2
Full length human recombit protein of human B3GALNT2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI1G2
Unconjugated
B3GALNT2
B3GalNAc-T2, MDDGA11
Mouse
Affinity Chromatography
RUO
148789
-20° C, Avoid Freeze/Thaw Cycles