Learn More
B3GALNT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17142-1-AP-150UL
This item is not returnable.
View return policy
Description
Beta-1,3-N-acetylgalactosaminyltransferase active in synthesizing a unique carbohydrate structure, GalNAc-beta-1-3GlcNAc, on N- and O-glycans. Has no galactose nor galactosaminyl transferase activity toward any acceptor substrate.Specifications
| B3GALNT2 | |
| Polyclonal | |
| Unconjugated | |
| B3GALNT2 | |
| B3GalNAc T2, B3GALNT2, Beta 1,3 GalNAc T2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 148789, 291212, 97884 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BG28, Q8NCR0 | |
| B3galnt2 | |
| B3GALNT2 Fusion Protein Ag10907 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |