Learn More
B3GAT2 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24631-1-AP-150UL
 View more versions of this product
Product Code. 18067045
  • € 410.00 / 150µL
Description
The B3GAT2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human B3GAT2. This antibody recognizes mouse, rat antigen. The B3GAT2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
B3GAT2
400 μg/mL
Rabbit
Primary
Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
B3GAT2, GlcAT D, GlcAT S, GLCATS, Glucuronosyltransferase S, KIAA1963, UDP glucuronosyltransferase S
150 μL
135152
Store at -20°C. Stable for one year after shipment.