Learn More
B3GAT2 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24631-1-AP-150UL
This item is not returnable.
View return policy
Description
The B3GAT2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human B3GAT2. This antibody recognizes mouse, rat antigen. The B3GAT2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| B3GAT2 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| B3GAT2, GlcAT D, GlcAT S, GLCATS, Glucuronosyltransferase S, KIAA1963, UDP glucuronosyltransferase S | |
| 150 μL | |
| 135152 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |