missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GNT2 Monoclonal Antibody (OTI1G10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505270S
This item is not returnable.
View return policy
Description
B3GNT2 Monoclonal antibody specifically detects B3GNT2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| B3GNT2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q05066 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6736 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1G10 | |
| Unconjugated | |
| SRY | |
| 3-Gn-T1; 3-Gn-T2; 3-N-a; B3GALT7; B3GNT; B3GNT1; B3GN-T1; B3GNT2; B3GN-T2; B3GNT-2; beta-1; Beta-1,3-galactosyltransferase 7; Beta-1,3-GalTase 7; beta-1,3-Gn-T1; beta-1,3-Gn-T2; Beta-1,3-N-acetylglucosaminyltransferase 1; beta-1,3-N-acetylglucosaminyltransferase bGnT-1; beta-1,3-N-acetylglucosaminyltransferase bGnT-2; BETA3GNT; beta3Gn-T1; beta3Gn-T2; Beta-3-Gx-T7; BGnT-1; BGNT2; BGnT-2; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 2; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 7; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 7; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2 | |
| Full length human recombinant protein of human SRY produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction