Learn More
B3GNT5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20422-1-AP-150UL
 View more versions of this product
Product Code. 16809964
  • € 410.00 / 150µL
Description
B3GNT5 is a member of the beta-1, 3-N-acetylglucosaminyltransferase family. This enzyme is a type II membrane protein. It exhibits strong activity to transfer GlcNAc to glycolipid substrates and is identified as the most likely candidate for lactotriaosylceramide synthase. This enzyme is essential for the expression of Lewis X epitopes on glycolipids.
Specifications
Specifications
B3GNT5
Polyclonal
Unconjugated
B3GNT5
B3GN T5, B3GNT5, Beta 1,3 Gn T5, beta3Gn T5, BGnT 5, Lactotriaosylceramide synthase, Lc(3)Cer synthase, Lc3 synthase
Rabbit
Antigen Affinity Chromatography
RUO
108105, 116740, 84002
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BGY6, Q99NB2, Q9BYG0
B3GNT5
B3GNT5 Fusion Protein Ag14246
150 μL
Primary
Human, Mouse, Rat