missing translation for 'onlineSavingsMsg'
Learn More

RCOR1/CoREST Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB600-240IR

Product Code. 18454113

  • € 681.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RCOR1/CoREST Polyclonal antibody specifically detects RCOR1/CoREST in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RCOR1/CoREST
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
KIAA0071CORESTRCORREST corepressor, Protein CoREST, REST corepressor 1
A synthetic peptide that maps to a region between residues 425 and the C-terminus (residue 482) of human REST (RE1-silencing transcription factor) corepressor using the numbering given in entry NP_055971.1 (GeneID 23186)
0.1 mL
Transcription Factors and Regulators
23186
Store at 4°C in the dark.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 755
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.