missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B3GNT9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562675
This item is not returnable.
View return policy
Description
Immunogen sequence: ELVVVHGLSA ADIWLMWRLL HGPHGPACAH PQPVAAGPFQ WDS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
B3GNT9 gene ontology annotations related to this gene include poly-N-acetyllactosamine biosynthetic process; protein glycosylation.
Specifications
| B3GNT9 | |
| Polyclonal | |
| Unconjugated | |
| B3GNT9 | |
| B3GNT9; beta-1,3-galactosyltransferase-related protein; beta-1,3-Gn-T9; beta-1,3-N-acetylglucosaminyltransferase 9; beta3Gn-T9; BGnT-9; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9; UNQ1922/PRO4397 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84752 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UX72 | |
| B3GNT9 | |
| Recombinant protein corresponding to Human B3GNT9. Recombinant protein control fragment (Product #RP-101710). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction