missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALT4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35457-20ul
This item is not returnable.
View return policy
Description
B4GALT4 Polyclonal antibody specifically detects B4GALT4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| B4GALT4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| B4Gal-T4, beta-1,4-galactosyltransferase 4, Beta-1,4-GalTase 4, beta4Gal-T4, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 4, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 4, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 4, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 35-344 of human B4GALT4 (NP_003769.1).,, Sequence:, VGAIQEIPKAKEFMANFHKTLILGKGKTLTNEASTKKVELDNCPSVSPYLRGQSKLIFKPDLTLEEVQAENPKVSRGRYRPQECKALQRVAILVPHRNREKHLMYLLEHLHPFLQRQQLDYGIYVIHQAEGKKFNRAKLLNVGYLEALKEENWDCFIFHDVDLVPENDFNLYKCEEHPKHLVVGRNSTGYRLRYSGYFGGVTALSREQFFKVNGFSNNYWGWGGEDDDLRLRVELQRMKISRPLPEVGKYTMVFHTRDKGNEVNAERMKLLHQVSRVWRTDGLSSCSYKLVSVEHNPLYINITVDFWFGA | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8702 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction