Learn More
B4GALT4 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30478-1-AP-150UL
 View more versions of this product
Product Code. 18009885
  • € 410.00 / 150µL
Description
The B4GALT4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human B4GALT4. This antibody recognizes human, mouse, rat antigen. The B4GALT4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
B4GALT4
400 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
B4Gal T4, B4GALT4, Beta 1,4 GalTase 4, beta4Gal T4
150 μL
8702
Store at -20°C. Stable for one year after shipment.