missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80 Antibody (016), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90956AF405
This item is not returnable.
View return policy
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Cynomolgus Monkey samples. It is validated for ELISA.
Specifications
| B7-1/CD80 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus B7-1/CD80 (; Met1-Asn242). | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
| 941 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 16 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction