missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80 Antibody (62N3G8), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25255APC
This item is not returnable.
View return policy
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| B7-1/CD80 | |
| Monoclonal | |
| APC | |
| NP_005182 | |
| CD80 | |
| Amino acids 102-223 of human CD80 were used as the immunogen for the antibody. | |
| 0.1 mL | |
| Adaptive Immunity, Cytokine Research, Immunology, Signal Transduction | |
| 941 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 62N3G8 | |
| Flow Cytometry | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Human. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction