missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80 Antibody (62N3G8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80588
This item is not returnable.
View return policy
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.
Specifications
| B7-1/CD80 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot reported by customer review, Flow Cytometry 1:10-1:1000. Use reported by customer review, Immunohistochemistry : 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunocytochemistry | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse reactivity reported in scientific literature (PMID: 27440778). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 62N3G8 | |
| Unconjugated | |
| PBS | |
| CD80 | |
| Amino acids 102-223 of human CD80 were used as the immunogen for the antibody. | |
| 0.1 mg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
| 941 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction