missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80 Antibody (C80/1608), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54307FR
This item is not returnable.
View return policy
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for ELISA, CyTOF
Specifications
| B7-1/CD80 | |
| Monoclonal | |
| DyLight 680 | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| Recombinant full-length human B7-1/CD80 protein (Uniprot: P33681) | |
| 100 μg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
| 941 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, CyTOF | |
| C80/1608 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction