missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-2/CD86 Antibody (BU63), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34569APCCY7
This item is not returnable.
View return policy
Description
B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Knockout Validated.
Specifications
| B7-2/CD86 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CD86 | |
| ARH-77 (B-lymphoblastoid cell line) | |
| 0.1 mL | |
| Primary | |
| Recognizes a protein of 70kDa, which is identified as CD86 (HLDA V; WS Code BP BP072. HLDA V; WS Code A A109. HLDA VI; WS Code BP 95. HLDA VI; WS Code B CD86.9). CD86 is a type I transmembrane glycoprotein and a member of the immunoglobulin superfamily of cell surface receptors. It is expressed at high levels on resting peripheral monocytes and dendritic cells and at very low density on resting B and T lymphocytes. CD86 expression is rapidly upregulated by B cell specific stimuli with peak expression at 18 to 42 hours after stimulation. CD86, along with CD80/B71, is an important accessory molecule in T cell co-stimulation via its interaction with CD28 and CD152/CTLA4. Since CD86 has rapid kinetics of induction, it is believed to be the major CD28 ligand expressed early in the immune response. It is also found on malignant Hodgkin and Reed Sternberg (HRS) cells in Hodgkin′s disease. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry, Immunoassay | |
| BU63 | |
| Flow Cytometry, Knockout Validated | |
| Activation B7-2 antigen, B70, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
| Mouse | |
| Protein G purified | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Dendritic Cell Markers, Diabetes Research, Immunology, Myeloid derived Suppressor Cell | |
| 942.0 | |
| Human, Mouse, Rat | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction