missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H2/ICOSLG Antibody (136726), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB165APCCY7
This item is not returnable.
View return policy
Description
B7-H2/ICOSLG Monoclonal antibody specifically detects B7-H2/ICOSLG in Human samples. It is validated for Flow Cytometry
Specifications
| B7-H2/ICOSLG | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 136726 | |
| Flow Cytometry | |
| B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
| Mouse myeloma cell line NS0-derived recombinant human B7-H2, Asp19-Ser258, Accession # O75144 | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23308 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction