Learn More
B7-H3/CD276 Antibody (185504), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1027MFV610
Product Code. 18511026
  • € 718.00 / 0.10mL
Description
B7-H3/CD276 Monoclonal antibody specifically detects B7-H3/CD276 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF, KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
B7-H3/CD276
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
Immunology
80381
Store at 4C in the dark.
Western Blot, Flow Cytometry, CyTOF, KnockDown
185504
Western Blot, Flow Cytometry, CyTOF-ready, Knockout Validated
B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule
Mouse myeloma cell line NS0-derived recombinant human B7-H3, Leu29-Pro245, Accession # NP_079516