missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H3/CD276 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32251F
This item is not returnable.
View return policy
Description
B7-H3/CD276 Polyclonal antibody specifically detects B7-H3/CD276 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| B7-H3/CD276 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule | |
| The immunogen this antibody was made to, maps to a region between residue 484 to 534 of human CD276 Molecule using the numbering given in entry NP_001019907.1 (GeneID 80381). | |
| 0.1 mL | |
| Immunology | |
| 80381 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction