missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H3/CD276 Antibody (MM0104-20J12), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12048MFV500
This item is not returnable.
View return policy
Description
B7-H3/CD276 Monoclonal antibody specifically detects B7-H3/CD276 in Human samples. It is validated for Western Blot, Immunocytochemistry
Specifications
| B7-H3/CD276 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| MM0104-20J12 | |
| Western Blot, Immunocytochemistry | |
| B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule | |
| Human recombinant B7-H3/CD276 (extracellular domain) | |
| 0.1 mL | |
| Immunology | |
| 80381 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction