missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H3/CD276 Antibody (omburtamab) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28510JF525
This item is not returnable.
View return policy
Description
B7-H3/CD276 Monoclonal antibody specifically detects B7-H3/CD276 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| B7-H3/CD276 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule | |
| B7-H3/ CD276 | |
| 0.1 mL | |
| Immunology | |
| 80381 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| omburtamab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction