missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B7-H3 Monoclonal Antibody (01)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530253
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human B7-H3/CD276.
CD276 is a member of the B7 family of co-stimulatory molecules also known as B7-H3. CD276 is a type I transmembrane protein that induces the proliferation of CD4+ and CD8+ T cells, enhances the generation of cytotoxic T cells and selectively stimulates the production of interferon gamma. Expression of CD276 can be induced on dendritic cells and monocytes by inflammatory cytokines, and is also widely expressed in peripheral tissues including the heart, kidney, testis and colon. In humans, CD276 exists as two isoforms which result from gene duplication and differential splicing. CD276 is reported to have therapeutic potential for the regulation of cell-mediated immune responses to cancer, particularly in conjunction with anti-angiogenic therapy.
Specifications
| B7-H3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q5ZPR3 | |
| CD276 | |
| Recombinant Human B7-H3/CD276 Protein (Met1-Thr461). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 01 | |
| Unconjugated | |
| CD276 | |
| 4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352 | |
| Mouse | |
| Protein A | |
| RUO | |
| 80381 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction