Learn More
Novus Biologicals™ B7-H4 Antibody (Millennium patent anti-B7-H4), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28513-100ug
 View more versions of this product
Product Code. 30046008
  • € 338.00 / 100µg
Description
B7-H4 Monoclonal antibody specifically detects B7-H4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
B7-H4
Monoclonal
Unconjugated
Q7Z7D3
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Millennium patent anti-B7-H4
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1
B7-H4/ VTCN1
100 μg
Cancer, Cell Cycle and Replication
79679