missing translation for 'onlineSavingsMsg'
Learn More

B7-H4 Antibody (PSH03-65), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-32065

Product Code. 30115859

  • € 508.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

B7-H4 Monoclonal antibody specifically detects B7-H4 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

B7-H4
Monoclonal
1 mg/mL
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1
Synthetic peptide within human B7-H4 aa 101-150/ 282. (Uniprot: Q7Z7D3)
100 μL
Cancer, Cell Cycle and Replication
79679
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunocytochemistry/Immunofluorescence
PSH03-65
Unconjugated
PBS (pH7.4), 0.1% BSA and 40% Glycerol
Rabbit
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.