missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H6 Antibody (1002910) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10162JF525
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Cynomolgus Monkey,Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| B7-H6 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| B7 homolog 6, B7-H6, DKFZp686O24166 | |
| Human embryonic kidney cell, HEK293-derived Cynomolgus Monkey B7-H6, Asp25-Ser262 | |
| 0.1 mL | |
| Cancer, Immunology | |
| 374383 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Western Blot, CyTOF | |
| 1002910 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Cynomolgus Monkey, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction