missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H6 Antibody (17B1.3) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-09050-0.025mg
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| B7-H6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| 17B1.3 | |
| Unconjugated | |
| B7 homolog 6, B7-H6, DKFZp686O24166 | |
| Immunised mice with extracellular domain of human B7-H6 and selected on the basis of specific reactivity by P815.B7-H6, but not with P815.B7-H1. | |
| 0.025 mg | |
| Cancer, Immunology | |
| 374383 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction