Learn More
B7-H6 Antibody (2167B), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB71443I
Product Code. 18531996
  • € 624.00 / 0.10mL
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
B7-H6
Monoclonal
Janelia Fluor 549
50mM Sodium Borate
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
2167B
Flow Cytometry, CyTOF-ready
B7 homolog 6, B7-H6, DKFZp686O24166
Human embryonic kidney cell, HEK293-derived human B7-H6 protein, Asp25-Ser262, Accession # Q68D85
0.1 mL
Cancer, Immunology
374383
Store at 4°C in the dark.