missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H6 Antibody (875001), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7144MFV450
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF, KnockDown
Specifications
| B7-H6 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Immunology | |
| 374383 | |
| Store at 4C in the dark. |
| Flow Cytometry, ELISA, CyTOF, KnockDown | |
| 875001 | |
| Flow Cytometry, ELISA, CyTOF-ready, Knockout Validated | |
| B7 homolog 6, B7-H6, DKFZp686O24166 | |
| Mouse myeloma cell line NS0-derived recombinant human B7-H6, Met1-Ser262, Accession # Q68D85 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction