Learn More
B7-H6 Antibody (875002) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB71442A
Product Code. 18612544
  • € 680.00 / 0.10mL
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
B7-H6
Monoclonal
APC
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, ELISA, CyTOF
875002
Flow Cytometry, ELISA, CyTOF-ready
B7 homolog 6, B7-H6, DKFZp686O24166
Mouse myeloma cell line NS0-derived recombinant human B7-H6, Met1-Ser262, Accession # Q68D85
0.1 mL
Cancer, Immunology
374383
Store at 4°C in the dark.