missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H6 Antibody (875023), Allophycocyanin/Cy7, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB71441APCCY7
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry, Product Image.
Specifications
| B7-H6 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| NCR3LG1 | |
| Partial recombinant human B7-H6 protein (amino acids1-262). [UniProt Q68D85] | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 875023 | |
| Flow Cytometry | |
| B7 homolog 6, B7-H6, DKFZp686O24166 | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Immunology | |
| 374383.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction