Learn More
B7-H6 Antibody (Dartmouth patent anti-B7-H6), Biotin, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-27971B
Product Code. 30116885
  • € 431.00 / 0.10mL
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
B7-H6
Monoclonal
Biotin
B7 homolog 6, B7-H6, DKFZp686O24166
B7-H6/ NCR3LG1
0.1 mL
Cancer, Immunology
374383
Store at 4°C in the dark.
IgG
Flow Cytometry, ELISA, Functional Assay
Dartmouth patent anti-B7-H6
PBS
Human
Protein A purified