missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H7/HHLA2 Antibody (907812R), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB80841RM
This item is not returnable.
View return policy
Description
B7-H7/HHLA2 Monoclonal antibody specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Specifications
| B7-H7/HHLA2 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 907812R | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
| Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2, Met1-Asn344, Accession # Q9UM44 | |
| 0.1 mL | |
| Immunology, Inflammation | |
| 11148 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction