missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B9D1 Polyclonal Antibody

Description
Immunogen sequence: ASPSVFLLMV NGQVESAQFP EYDDLYCKYC FVYGQDWAPT AGLEEGISQI TSKSQDVRQA LVWN Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 98%.
This gene encodes a B9 domain-containing protein, one of several that are involved in ciliogenesis. Alterations in expression of this gene have been found in a family with Meckel syndrome. Meckel syndrome has been associated with at least six different genes. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Three alternatively spliced transcript variants that encode different proteins have been described for this gene.
Specifications
Specifications
| Antigen | B9D1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | B9D1 |
| Gene Accession No. | Q9UPM9 |
| Gene Alias | AW045994; B9; B9 domain containing 1; B9 domain-containing protein 1; B9 protein domain 1; B9D1; Endothelial precursor cells protein B9; endothelial precursor protein B9; Eppb9; MKS1-related protein 1; MKS9; MKSR1 |
| Gene Symbols | B9D1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?