missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BA Antigen (Leu21) Antibody (DF1588), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54564IR
This item is not returnable.
View return policy
Description
BA Antigen (Leu21) Monoclonal antibody specifically detects BA Antigen (Leu21) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
Specifications
| BA Antigen (Leu21) | |
| Monoclonal | |
| DyLight 755 | |
| CD78, CD78, Cdw78, Cdw78, LO panB a | |
| Human leucocytes | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, CyTOF | |
| DF1588 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction