missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572656
This item is not returnable.
View return policy
Description
BA2 Polyclonal Antibody for Western Blot
Involved in oxygen transport from gills to the various peripheral tissues.
Specifications
| BA2 | |
| Polyclonal | |
| Unconjugated | |
| ba2 | |
| ba2; Beta-2-globin; Beta-A2-globin | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 30217 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.81 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q90485 | |
| ba2 | |
| KLH conjugated synthetic peptide between 71-104 amino acids of DANRE ba2. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction