missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAAT Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92503-0.02ml
This item is not returnable.
View return policy
Description
BAAT Polyclonal antibody specifically detects BAAT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| BAAT | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| BACAT, BATbile acid-CoA:amino acid N-acyltransferase, bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase), bile acid Coenzyme A: amino acid N-acyltransferase (glycineN-choloyltransferase), EC 2.3.1.65, EC 3.1.2.2, FLJ20300, Glycine N-choloyltransferase, Long-chain fatty-acyl-CoA hydrolase, MGC104432 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 309-418 of human BAAT (NP_001692.1). YLFPIEEAQGQFLFIVGEGDKTINSKAHAEQAIGQLKRHGKNNWTLLSYPGAGHLIEPPYSPLCCASTTHDLRLHWGGEVIPHAAAQEHAWKEIQRFLRKHLIPDVTSQL | |
| 0.02 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 570 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction