missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACE-1 Antibody (137612) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB931W
This item is not returnable.
View return policy
Description
BACE-1 Monoclonal antibody specifically detects BACE-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| BACE-1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 137612 | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2 | |
| Mouse myeloma cell line NS0-derived recombinant human BACE-1, Thr22-Tyr460, Accession # P56817 | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 23621 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction