Learn More
CLEC-2A Antibody (705518), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB7219H
Product Code. 18633495
  • ONLINE EXCLUSIVE PRICE
    € 608.00
    € 574.35 / 0.10mL
    Save € 33.65
    5% Off
Description
CLEC-2A Monoclonal antibody specifically detects CLEC-2A in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CLEC-2A
Monoclonal
HRP
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
387836
Store at 4C in the dark.
IgG1
Flow Cytometry, CyTOF
705518
Flow Cytometry, CyTOF-ready
CLEC2A C-type lectin domain family 2, member A, KACL, PILAR, UNQ5792
Mouse myeloma cell line NS0-derived recombinant human CLEC-2A, Trp49-Leu174 (predicted), Accession # Q6UVW9
0.1 mL