missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACE-1 Antibody (SR2142), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | BACE-1 |
|---|---|
| Clone | SR2142 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:40 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693744
|
Novus Biologicals
NBP3-21741-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671994
|
Novus Biologicals
NBP3-21741-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BACE-1 Monoclonal antibody specifically detects BACE-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| BACE-1 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:40 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2 | |
| A synthesized peptide derived from human BACE-1 (Uniprot #: P56817) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2142 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 23621 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title