missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACH1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71814UV
This item is not returnable.
View return policy
Description
BACH1 Polyclonal antibody specifically detects BACH1 in Human samples. It is validated for Western Blot
Specifications
| BACH1 | |
| Polyclonal | |
| Western Blot | |
| basic region leucine zipper transcriptional regulator BACH1, BTB and CNC homolog 1, BTB and CNC homology 1, basic leucine zipper transcription factor 1, HA2303, transcription regulator protein BACH1, transcription regulator protein10BACH-1 | |
| The immunogen for this product maps to a region between residue 350 and 400 of human BTB and CNC Homology 1, Basic Leucine Zipper Transcription Factor 1 using the numbering given in entry NP_001177.1 (GeneID 571). | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Repair | |
| 571 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?