missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BACH1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA14242

Product Code. 11588790

  • € 530.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PA1-4242 detects BACH1 from human samples. PA1-4242 has been successfully used in Western blot procedures. PA1-4242 immunizing peptide corresponds to amino acid residues 1233-1249 from human BACH1 protein.

BACH1.3 is a transcription factor that belongs to the cap'n'collar type of basic region leucine zipper factor family (CNC-bZip). The encoded protein contains broad complex, tramtrack, bric-a-brac/poxvirus and zinc finger (BTB/POZ) domains, which is atypical of CNC-bZip family members. These BTB/POZ domains facilitate protein-protein interactions and formation of homo and/or hetero oligomers. When this encoded protein forms a heterodimer with MafK, it functions as a repressor of Maf recognition element (MARE) and transcription is repressed. Multiple alternatively spliced transcript variants have been identified for this gene. Some exons of this gene overlap with some exons from the C21orf41 gene, which is transcribed in an opposite orientation to this gene, but which does not seem to encode a protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BACH1
Polyclonal
Unconjugated
BACH1
6230421P05Rik; AI323795; BACH1; BACH-1; basic region leucine zipper transcriptional regulator BACH1; BTB and CNC homolog 1; BTB and CNC homology 1; BTB and CNC homology 1 transcription factor; BTB and CNC homology 1, basic leucine zipper transcription factor 1; BTB domain and CNC homolog 1; BTBD24; HA2303; Transcription regulator protein BACH1
Rabbit
Antigen affinity chromatography
RUO
571
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O14867
BACH1
Synthetic Peptide: N(1233) F K P S P S K N K G M F P G F K(1249).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.